missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-hydroxybenzoic acid, TRC

Product Code. 30371579
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371579 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371579 Supplier Toronto Research Chemicals Supplier No. TRCD44742825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 74799-63-6
Molecular Formula C7H4F2O3
Quantity 25 mg
InChI Formula InChI=1S/C7H4F2O3/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,10H,(H,11,12)
SMILES OC(=O)c1cc(F)c(O)c(F)c1
Molecular Weight (g/mol) 174.1
Formula Weight 174.0129
Chemical Name or Material 3,5-Difluoro-4-hydroxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.