missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11368866
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368866 5 g 5g
11358866 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368866 Supplier Thermo Scientific Alfa Aesar Supplier No. L19507.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210532-25-5
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.59
MDL Number MFCD02091378
InChI Key IIQKIICAOXAXEJ-UHFFFAOYSA-N
Synonym 3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
PubChem CID 2774058
IUPAC Name 3,5-difluorobenzenesulfonyl chloride
SMILES FC1=CC(=CC(F)=C1)S(Cl)(=O)=O

Specifications

Melting Point 57°C to 59°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 8053561
Sensitivity Moisture sensitive
Formula Weight 212.6
Percent Purity 97%
Chemical Name or Material 3,5-Difluorobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.