missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 10014017 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10014017 5 g 5g
10084247 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 10014017

Brand: Thermo Scientific Acros 431000050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210532-25-5
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.59
MDL Number MFCD02091378
InChI Key IIQKIICAOXAXEJ-UHFFFAOYSA-N
Synonym 3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
PubChem CID 2774058
IUPAC Name 3,5-difluorobenzenesulfonyl chloride
SMILES FC1=CC(=CC(F)=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 55.9°C to 58.2°C
Color Orange to Tan
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 212.6
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Difluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Difluorobenzenesulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.