missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Difluoropropiophenone, 97%, Thermo Scientific™

Product Code. 11460691
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11460691

Brand: Thermo Scientific Alfa Aesar B20150.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135306-45-5
Molecular Formula C9H8F2O
Molecular Weight (g/mol) 170.159
MDL Number MFCD00061142
InChI Key FVDQWXARVQADKN-UHFFFAOYSA-N
Synonym 1-3,5-difluorophenyl propan-1-one, 3,5-difluoropropiophenone, 3',5'-difluoropropiophenone, acmc-1cehk, 1-3,5-difluorophenyl-1-propanone, 1-propanone,1-3,5-difluorophenyl, 1-3,5-difluoro-phenyl-propan-1-one, 1-3,5-bis fluoranyl phenyl propan-1-one
PubChem CID 145620
IUPAC Name 1-(3,5-difluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC(=C1)F)F

Specifications

Melting Point 25°C to 27°C
Boiling Point 82°C (10 mmHg)
Quantity 1 g
Beilstein 7422838
Formula Weight 170.16
Percent Purity 97%
Chemical Name or Material 3',5'-Difluoropropiophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.