missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dihydroxyacetophenone, 97%

Product Code. 10656612 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656612 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10656612 Supplier Thermo Scientific Acros Supplier No. 208630250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51863-60-6 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51863-60-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 143.0°C to 147.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H8O3
Linear Formula (HO)2C6H3COCH3
MDL Number MFCD00002290
Quantity 25 g
Beilstein 08, II, 301
Synonym 3',5'-dihydroxyacetophenone, 1-3,5-dihydroxyphenyl ethanone, 3,5-dihydroxyacetophenone, 5-acetylresorcinol, 1-3,5-dihydroxyphenyl ethan-1-one, unii-kp7r433nnr, ethanone, 1-3,5-dihydroxyphenyl, kp7r433nnr, 1-acetyl-3,5-dihydroxybenzene, 1-3,5-dihydroxyphenyl-ethanone
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in ethanol,slightly soluble in acetone
InChI Key WQXWIKCZNIGMAP-UHFFFAOYSA-N
SMILES CC(=O)C1=CC(=CC(=C1)O)O
IUPAC Name 1-(3,5-dihydroxyphenyl)ethanone
Molecular Weight (g/mol) 152.15
PubChem CID 103993
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3', 5'-Dihydroxyacetophenone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3',5'-Dihydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.