missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzoic Acid, TRC

Product Code. 30134563
Change view
Click to view available options
Quantity:
50 mg
50 g
100 g
Unit Size:
100g
50g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134563 100 g 100g
30133333 50 g 50g
30008875 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134563 Supplier Toronto Research Chemicals Supplier No. TRCD451700100G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-10-5
Molecular Formula C7 H6 O4
Molecular Weight (g/mol) 154.12
Synonym 3,5-Dihydroxybenzoic Acid, Terbutaline Sulphate Imp. A (EP), alpha-Resorcylic Acid, α-Resorcylic acid, 3,5-Dihydroxybenzoic acid, 5-Carboxyresorcinol, NSC 22948
IUPAC Name 3,5-dihydroxybenzoic acid
SMILES OC(=O)c1cc(O)cc(O)c1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C7H6O4/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,8-9H,(H,10,11)
Formula Weight 154.0266
Percent Purity >95%
Chemical Name or Material alpha-Resorcylic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.