missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylic Acid, TRC

Product Code. 30384943
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30384943

Brand: Toronto Research Chemicals TRCD455145100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Diiodosalicylic Acid
CAS 133-91-5
Molecular Formula C7H4I2O3
Quantity 100 g
Synonym 2-Hydroxy-3,5-diiodo-benzoic Acid, 2-Hydroxy-3,5-diiodobenzenecarboxylic acid, 2-Hydroxy-3,5-diiodobenzoic Acid, 3,5-Diiodo-2-hydroxybenzoic Acid, 3,5-Diiodosalicylic Acid, NSC 6303
InChI Formula InChI=1S/C7H4I2O3/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2,10H,(H,11,12)
SMILES OC(=O)c1cc(I)cc(I)c1O
Molecular Weight (g/mol) 389.91
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.