missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethoxytoluene, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 3,5-Dimethoxytoluene |
| CAS | 4179-19-5 |
| Molecular Formula | C9H12O2 |
| Quantity | 25 g |
| Synonym | 1,3-Dimethoxy-5-methylbenzene, 1-Methyl-3,5-dimethoxybenzene, 5-Methyl-1,3-dimethoxybenzene, 5-Methylresorcinol Dimethyl Ether, NSC 72352, Orcinol Dimethyl Ether |
| InChI Formula | InChI=1S/C9H12O2/c1-7-4-8(10-2)6-9(5-7)11-3/h4-6H,1-3H3 |
| SMILES | COc1cc(C)cc(OC)c1 |
| Molecular Weight (g/mol) | 152.19 |
| Formula Weight | 152.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction