missing translation for 'onlineSavingsMsg'
Learn More

3,5,-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1h-pyrazole, 97%, Thermo Scientific™

Product Code. 10264404
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10264404 1 g Amber Glass Bottle 1g
10471504 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10264404 Supplier Thermo Scientific Maybridge Supplier No. CC68239DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 937796-06-0
Molecular Formula C17H23BN2O2
Molecular Weight (g/mol) 298.19
MDL Number MFCD09879979
InChI Key SERWCEBXGSSDRB-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 4-3,5-dimethylpyrazol-1-yl phenylboronic acid, pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3,5,-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 4-3,5-dimethyl-1h-pyrazol-1-yl benzeneboronic acid,pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole
PubChem CID 24229763
IUPAC Name 3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
SMILES CC1=NN(C(C)=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 298.19
Percent Purity 97%
Chemical Name or Material 3,5,-Dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.