missing translation for 'onlineSavingsMsg'
Learn More

3,5,-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1h-pyrazole, 97%, Thermo Scientific™

Produktkod. 10471504
Change view
Klicka för att se tillgängliga alternativ
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging Förpackningsstorlek
10471504 250 mg Amber Glass Bottle 250mg
10264404 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10471504 Leverantör Thermo Scientific Maybridge Leverantörsnummer CC68239CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 937796-06-0
Molecular Formula C17H23BN2O2
Molecular Weight (g/mol) 298.19
MDL Number MFCD09879979
InChI Key SERWCEBXGSSDRB-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 4-3,5-dimethylpyrazol-1-yl phenylboronic acid, pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3,5,-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 4-3,5-dimethyl-1h-pyrazol-1-yl benzeneboronic acid,pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole
PubChem CID 24229763
IUPAC Name 3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
SMILES CC1=NN(C(C)=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 298.19
Percent Purity 97%
Chemical Name or Material 3,5,-Dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.