missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde, 99%, Thermo Scientific™

Product Code. 10062011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10062011 5 g Glass bottle 5g
10131231 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062011 Supplier Thermo Scientific Acros Supplier No. 408520050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 22042-79-1
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD00051396
InChI Key VOMRTQQGXWPTJK-UHFFFAOYSA-N
PubChem CID 89179
IUPAC Name 3,5-dimethyl-1-phenylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C2=CC=CC=C2)C)C=O

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125°C to 128°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 3,5-dimethyl-1-phenylpyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 3,5-dimethyl-1-phenyl, 3,5-dimethyl-1-phenyl-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3,5-dimethyl-1-phenyl, 3,5-dimethyl-4-formyl-1-phenylpyrazole, 3,5-dimethyl-1-phenyl-4-pyrazolecarboxaldehyde, 3?5-dimethyl-1-phenylpyrazole-4-carboxaldehyde, 3,5-dimethyl-1-phenyl 1h-pyrazole-4-carbaldehyde, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbaldehyde #
Formula Weight 200.24
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde, 99%
compliance-icons
Nombre del producto
  • 3,5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.