missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrazole-1-acetic acid, 95%, Thermo Scientific™

Product Code. 15505946 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15505946

Brand: Thermo Scientific Acros 456952500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16034-49-4
Molecular Formula C7H9N2O2
Molecular Weight (g/mol) 153.16
MDL Number MFCD00297249
InChI Key JYSWEDYPQJOEPO-UHFFFAOYSA-M
Synonym 2-3,5-dimethyl-1h-pyrazol-1-yl acetic acid, 3,5-dimethyl-pyrazol-1-yl-acetic acid, 3,5-dimethyl-1h-pyrazol-1-yl acetic acid, 3,5-dimethylpyrazol-1-yl acetic acid, 1h-pyrazole-1-acetic acid, 3,5-dimethyl, 3-nitro-benzene-thiol, 3,5-dimethyl-1h-pyrazole-1-acetic acid, 1h-pyrazole-1-aceticacid, 3,5-dimethyl, 2-3,5-dimethylpyrazolyl acetic acid, 3,5-dimethyl-pyrazol-1-yl acetic acid
PubChem CID 572875
IUPAC Name 2-(3,5-dimethylpyrazol-1-yl)acetic acid
SMILES CC1=NN(CC([O-])=O)C(C)=C1

Specifications

Melting Point 186.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 154.17
Percent Purity 95%
Chemical Name or Material 3,5-Dimethyl-1H-pyrazole-1-acetic acid
compliance-icons
Product Identifier
  • 3,5-Dimethyl-1H-pyrazole-1-acetic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.