missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10792663
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10792663 1 g 1g
10374612 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10792663 Supplier Thermo Scientific Maybridge Supplier No. CC01201DA
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 113808-86-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00159642
InChI Key IOOWDXMXZBYKLR-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl, 1h-pyrazole-4-carboxylicacid, 3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-carboxylicacid, acmc-20a7j9, pyrazole carboxylic acid 1, ksc173i7f, 3,5-dimethylpyrazole-4-carboxylic, 3,5-dimethylpyrazole-4-carboxylicacid, 1h-pyrazole-4-carboxylicacid,3,5-dimethyl
PubChem CID 2776164
IUPAC Name 3,5-dimethyl-1H-pyrazole-4-carboxylic acid
SMILES CC1=C(C(=NN1)C)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.