missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-isoxazolecarbaldehyde, Thermo Scientific™

Product Code. 10623795
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10623795 10 g 10g
10437802 250 mg 250mg
10384182 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10623795 Supplier Thermo Scientific Maybridge Supplier No. CC00604EA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 54593-26-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
InChI Key TVAYXKLCEILMEA-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-isoxazolecarbaldehyde, 3,5-dimethylisoxazole-4-carbaldehyde, 3,5-dimethylisoxazole-4-carboxaldehyde, 4-isoxazolecarboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-4-isoxazolecarboxaldehyde, 3,5-dimethyl-4-formalisoxazole, pubchem8682, acmc-1axd1, ksc496e3j, 3,5-dimethylisoxazol-4-carbaldehyde
PubChem CID 289576
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbaldehyde
SMILES CC1=C(C(=NO1)C)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-4-isoxazolecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.