missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-methoxyphenylboronic acid, 95%, Thermo Scientific™

Product Code. 11916031 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916031 1 g 1g
11926031 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916031 Supplier Thermo Scientific Acros Supplier No. 444030010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 301699-39-8
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
InChI Key WZUCSPWZHRVOSD-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-methoxyphenylboronic acid, 3,5-dimethyl-4-methoxybenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boronic acid, 4-methoxy-3,5-dimethyl-phenyl boronic acid, 4-methoxy-3,5-dimethylbenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boranediol, boronic acid, 4-methoxy-3,5-dimethylphenyl, pubchem6937, acmc-209hdl, 3,5-dimethyl-4-methoxybenzeneboronicacid
PubChem CID 3817586

Specifications

Melting Point 203°C to 204°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 180.01
Percent Purity 95%
Chemical Name or Material 3, 5-Dimethyl-4-methoxyphenylboronic acid
compliance-icons
Product Identifier
  • 3,5-Dimethyl-4-methoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.