missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Melting Point | 9.8°C |
| CAS Min % | 97.5 |
| Color | Brown to Brown-Yellow |
| CAS Max % | 100.0 |
| Density | 0.9700g/mL |
| Quantity | 100 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Refractive Index | 1.5560 to 1.558 |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- 3,5-Dimethylaniline
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
- H411-Toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?