missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylaniline, TRC

Product Code. 30367351
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30367351 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30367351

Brand: Toronto Research Chemicals TRCD236306250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Xylidine
CAS 108-69-0
Molecular Formula C8 H11 N
Quantity 250 mg
Synonym 3,5-Xylidine, 3,5-Dimethylbenzenamine, 3,5-Dimethyl-1-aminobenzene, 3,5-Dimethyl-N-phenylamine, 3,5-Dimethylaniline, 3,5-Dimethylphenylamine, 3,5-Xylylamine, 5-Amino-1,3-dimethylbenzene, 5-Amino-1,3-xylene, NSC 26880
InChI Formula InChI=1S/C8H11N/c1-6-3-7(2)5-8(9)4-6/h3-5H,9H2,1-2H3
SMILES Cc1cc(C)cc(N)c1
IUPAC Name 3,5-dimethylaniline
Molecular Weight (g/mol) 121.18
Formula Weight 121.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.