missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylaniline, TRC

Product Code. 30363277
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30363277 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30363277

Brand: Toronto Research Chemicals TRCD2363062.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Xylidine
CAS 108-69-0
Molecular Formula C8 H11 N
Quantity 2.5 g
Synonym 3,5-Xylidine, 3,5-Dimethylbenzenamine, 3,5-Dimethyl-1-aminobenzene, 3,5-Dimethyl-N-phenylamine, 3,5-Dimethylaniline, 3,5-Dimethylphenylamine, 3,5-Xylylamine, 5-Amino-1,3-dimethylbenzene, 5-Amino-1,3-xylene, NSC 26880
InChI Formula InChI=1S/C8H11N/c1-6-3-7(2)5-8(9)4-6/h3-5H,9H2,1-2H3
SMILES Cc1cc(C)cc(N)c1
IUPAC Name 3,5-dimethylaniline
Molecular Weight (g/mol) 121.18
Formula Weight 121.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.