missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylanisole, 99%

Product Code. 11499087
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Pack of 144
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499087 25 g 25g
11754199 25 g Pack of 144
11409097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499087 Supplier Thermo Scientific Alfa Aesar Supplier No. B20651.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 874-63-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-63-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008398
InChI Key JCHJBEZBHANKGA-UHFFFAOYSA-N
Synonym 3,5-dimethylanisole, 5-methoxy-m-xylene, benzene, 1-methoxy-3,5-dimethyl, 1-methoxy-3,5-dimethyl-benzene, 3,5-dimethylanisol, 3,5-dimethylanizole, acmc-1bkxt, anisole, 3,5-dimethyl, ksc494e7h, 5-methoxy-1,3-dimethylbenzene
PubChem CID 70126
IUPAC Name 1-methoxy-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)OC)C

Specifications

Density 0.96
Boiling Point 192°C to 193°C
Quantity 25 g
Refractive Index 1.513
Beilstein 2040905
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3,5-Dimethylanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.