missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-boronic acid, pinacol ester, Thermo Scientific™

Product Code. 10638350
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10638350 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10638350 Supplier Thermo Scientific Maybridge Supplier No. MAY00023DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 832114-00-8
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H18BNO3
Quantity 5 g
Synonym 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole, 3,5-dimethylisoxazole-4-boronic acid pinacol ester, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl isoxazole, 3,5-dimethylisoxazole-4-boronic acid, pinacol ester, 3,5-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 3,5-dimethyl-4-isoxazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3,5-dimethylisoxazol-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pubchem18439, acmc-209prk
InChI Key CVLHETBAROWASE-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(ON=C2C)C
IUPAC Name 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
Molecular Weight (g/mol) 223.079
PubChem CID 2758656
Formula Weight 223.08
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethylisoxazole-4-boronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.