missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carbonyl chloride, 98%, Thermo Scientific™

Product Code. 11450250
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450250 1 g 1g
11460250 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450250 Supplier Thermo Scientific Alfa Aesar Supplier No. H50695.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dimethylisoxazole-4-carbonyl chloride is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with bases, oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31301-45-8
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.569
MDL Number MFCD00052555
InChI Key MPYGFFPGJMGVSW-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-carbonyl chloride, 4-isoxazolecarbonyl chloride, 3,5-dimethyl, 3,5-dimethylisoxazole-4-carbonylchloride, dimethyl-1,2-oxazole-4-carbonyl chloride, 3,5-dimethyl-4-isoxazolecarbonyl chloride, pubchem8669, acmc-20a3wc, 3,5-dimethylisoxasole-4-carbonyl chloride, 3,5-dimethyisoxazole-4-carbonyl chloride, 3,5 dimethylisoxazole-4-carbonyl chloride
PubChem CID 2736265
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C)C(=O)Cl

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 159.57
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylisoxazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.