missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carbonyl chloride, Thermo Scientific™

Product Code. 10603783
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10603783 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10603783 Supplier Thermo Scientific Maybridge Supplier No. CC00602DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 31301-45-8
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C6H6ClNO2
MDL Number MFCD00052555
Quantity 1 g
Synonym 3,5-dimethylisoxazole-4-carbonyl chloride, 4-isoxazolecarbonyl chloride, 3,5-dimethyl, 3,5-dimethylisoxazole-4-carbonylchloride, dimethyl-1,2-oxazole-4-carbonyl chloride, 3,5-dimethyl-4-isoxazolecarbonyl chloride, pubchem8669, acmc-20a3wc, 3,5-dimethylisoxasole-4-carbonyl chloride, 3,5-dimethyisoxazole-4-carbonyl chloride, 3,5 dimethylisoxazole-4-carbonyl chloride
InChI Key MPYGFFPGJMGVSW-UHFFFAOYSA-N
SMILES CC1=C(C(=NO1)C)C(=O)Cl
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbonyl chloride
Molecular Weight (g/mol) 159.569
PubChem CID 2736265
Formula Weight 159.57
Percent Purity 95%
Chemical Name or Material 3, 5-Dimethylisoxazole-4-carbonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.