missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxaldehyde, 97%

Product Code. 15438508
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438508 25 g 25g
15428508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438508 Supplier Thermo Scientific Alfa Aesar Supplier No. H61912.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54593-26-9 | C6H7NO2 | 125.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54593-26-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD02681977
InChI Key TVAYXKLCEILMEA-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-isoxazolecarbaldehyde, 3,5-dimethylisoxazole-4-carbaldehyde, 3,5-dimethylisoxazole-4-carboxaldehyde, 4-isoxazolecarboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-4-isoxazolecarboxaldehyde, 3,5-dimethyl-4-formalisoxazole, pubchem8682, acmc-1axd1, ksc496e3j, 3,5-dimethylisoxazol-4-carbaldehyde
PubChem CID 289576
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbaldehyde
SMILES CC1=C(C(=NO1)C)C=O

Specifications

Melting Point 35°C to 38°C
Quantity 25 g
Formula Weight 125.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,5-Dimethylisoxazole-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.