missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole, 99%

Product Code. 11414893
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414893 25 g 25g
11424893 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414893 Supplier Thermo Scientific Alfa Aesar Supplier No. A12217.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 300-87-8 | C5H7NO | 97.117 g/mol

The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-87-8
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.117
MDL Number MFCD00003156
InChI Key FICAQKBMCKEFDI-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole, isoxazole, 3,5-dimethyl, 3,5-dimethylisooxazole, 3,5-dwumetyloizoksazolu, 3,5-dwumetyloizoksazolu polish, 3,5-dimethylisoxazol, pubchem8641, isoxazole,5-dimethyl, 3,5-dimethyl-isoxazole
PubChem CID 9312
IUPAC Name 3,5-dimethyl-1,2-oxazole
SMILES CC1=CC(=NO1)C

Specifications

Melting Point -14°C
Density 0.992
Boiling Point 142°C to 144°C
Flash Point 31°C (87°F)
Refractive Index 1.4415
Quantity 25 g
UN Number UN1993
Beilstein 106324
Solubility Information Insoluble in water.
Formula Weight 97.12
Percent Purity 99%
Chemical Name or Material 3,5-Dimethylisoxazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.