missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenol, TRC

Product Code. 30452909
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452909 100 g 100g
30450220 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452909 Supplier Toronto Research Chemicals Supplier No. TRCX749505100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 108-68-9
Molecular Formula C8 H10 O
Molecular Weight (g/mol) 122.16
Synonym 3,5-Dimethylphenol, Phenol, 3,5-dimethyl-, 3,5-Xylenol (8CI), 3,5-Dimethylphenol, 1,3,5-Xylenol, 1-Hydroxy-3,5-dimethylbenzene, NSC 9268
IUPAC Name 3,5-dimethylphenol
SMILES Cc1cc(C)cc(O)c1

Specifikationer

Quantity 100 g
InChI Formula InChI=1S/C8H10O/c1-6-3-7(2)5-8(9)4-6/h3-5,9H,1-2H3
Formula Weight 122.0732
Chemical Name or Material 3,5-Dimethylphenol
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.