missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenyl isocyanate, 97%

Product Code. 11322577
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322577 5 g 5g
11312577 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322577 Supplier Thermo Scientific Alfa Aesar Supplier No. L11698.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54132-75-1 | C9H9NO | 147.177 g/mol

3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54132-75-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013868
InChI Key DZSGDHNHQAJZCO-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl isocyanate, 3,5-dimethylphenylisocyanate, 3,5-xylyl isocyanate, benzene, 1-isocyanato-3,5-dimethyl, isocyanic acid 3,5-dimethylphenyl ester, 5-isocyanato-m-xylene, 1-isocyanato-3,5-dimethyl-benzene, acmc-1axct, 3,5-dimethylbenzenisocyanate, ksc274i2t
PubChem CID 521488
IUPAC Name 1-isocyanato-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)N=C=O)C

Specifications

Density 1.045
Boiling Point 205°C
Quantity 5 g
Refractive Index 1.528
UN Number UN2206
Beilstein 774958
Solubility Information Hydrolyzes in water.
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.