missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenyl isothiocyanate, 99%

Product Code. 11473684
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11473684 5 g 5g
11483684 25 g 25g
11493684 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11473684

Brand: Thermo Scientific Alfa Aesar A11302.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 40046-30-8 | C9H9NS | 163.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40046-30-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00041082
InChI Key DSMXCADWIFIJEX-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
PubChem CID 142406
IUPAC Name 1-isothiocyanato-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)N=C=S)C

Specifications

Quantity 5 g
UN Number UN2922
Beilstein 6856812
Formula Weight 163.24
Percent Purity 99%
Chemical Name or Material 3,5-Dimethylphenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.