missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylacetic acid, 98+%

Product Code. 11392217
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392217 1 g 1g
11302227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392217 Supplier Thermo Scientific Alfa Aesar Supplier No. L09707.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42288-46-0 | C10H11O2 | 163.20 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42288-46-0
Molecular Formula C10H11O2
Molecular Weight (g/mol) 163.20
MDL Number MFCD00082776
InChI Key HDNBKTWQBJJYPD-UHFFFAOYSA-M
Synonym 3,5-dimethylphenylacetic acid, 2-3,5-dimethylphenyl acetic acid, 3,5-dimethylphenyl acetic acid, benzeneacetic acid, 3,5-dimethyl, 3,5-xylyl acetic acid, acmc-20anc9, ksc238q4h, 3,5-dimethylbenzeneacetic acid, 3,5-dimethyl-phenyl-acetic acid
PubChem CID 4749495
IUPAC Name 2-(3,5-dimethylphenyl)acetic acid
SMILES CC1=CC(CC([O-])=O)=CC(C)=C1

Specifications

Melting Point 99°C to 103°C
Quantity 1 g
Beilstein 2083903
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.