missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylhydrazine hydrochloride, 95%, BLD Pharm™

Product Code. 31098773
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31098773 500 g
31105210 10 g
31095964 25 g
31100543 100 g
31098156 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31098773 Supplier BLD Pharm Supplier No. BD10694500g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 60481-36-9
Molecular Formula C8H13ClN2
MDL Number MFCD00052269
Quantity 500 g
SMILES Cl.CC1=CC(=CC(=C1)NN)C
Molecular Weight (g/mol) 172.66
Percent Purity 95%
Chemical Name or Material 3,5-Dimethylphenylhydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.