missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoyl chloride, 98+%

Product Code. 11432738
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432738 25 g 25g
11442738 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432738 Supplier Thermo Scientific Alfa Aesar Supplier No. A16729.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-33-2 | C7H3ClN2O5 | 230.56 g/mol

Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-33-2
Molecular Formula C7H3ClN2O5
Molecular Weight (g/mol) 230.56
MDL Number MFCD00007248
InChI Key NNOHXABAQAGKRZ-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
PubChem CID 7432
IUPAC Name 3,5-dinitrobenzoyl chloride
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl

Specifications

Melting Point 67°C to 70°C
Boiling Point 196°C (12 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 990249
Sensitivity Moisture sensitive
Merck Index 14,3277
Solubility Information Slightly soluble in water.
Formula Weight 230.57
Percent Purity ≥98%
Chemical Name or Material 3,5-Dinitrobenzoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Dinitrobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: 3,5-dinitro-benzoylchloride

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.