missing translation for 'onlineSavingsMsg'
Learn More

3(5)-Phenyl-1H-pyrazole, 99+%

Product Code. 10559623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10559623 10 g Glass bottle 10g
10572841 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10559623 Supplier Thermo Scientific Acros Supplier No. 296030100
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2458-26-6 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2458-26-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00159654
InChI Key OEDUIFSDODUDRK-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
PubChem CID 17155
IUPAC Name 5-phenyl-1H-pyrazole
SMILES N1N=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 75.0°C to 77.0°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 10 g
Beilstein 23, IV, 1238
Formula Weight 144.18
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 3(5)-Phenyl-1H-pyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 3(5)-Phenyl-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.