missing translation for 'onlineSavingsMsg'
Learn More

3-[5-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl]propan-1-ol, 97%, Thermo Scientific™

Product Code. 10445492
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10445492 10 g Amber Glass Bottle 10g
10394362 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10445492 Supplier Thermo Scientific Maybridge Supplier No. BTB05684EA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-15-6
Molecular Formula C11H11F3N2O
Molecular Weight (g/mol) 244.217
MDL Number MFCD00067735
InChI Key FDRMNTVCDKSRNL-UHFFFAOYSA-N
Synonym 3-5-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol, 2-3-hydroxy-1-propyl-5-trifluoromethyl benzimidazole, 3-5-trifluoromethyl-1h-1,3-benzodiazol-2-yl propan-1-ol, 2-3-hydroxy-n-propyl-5-trifluoromethyl-benzimidazole, 3-6-trifluoromethyl-1h-benzimidazol-2-yl propan-1-ol, 2-3-hydroxypropyl-5-trifluoromethyl benzimidazole, 2-3-hydroxy-n-propyl-5-trifluoromethyl benzimidazole, 2-3-hydroxypropyl-5-trifluoromethyl-1h-benzimidazole, 3-6-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol
PubChem CID 2775098
IUPAC Name 3-[6-(trifluoromethyl)-1H-benzimidazol-2-yl]propan-1-ol
SMILES C1=CC2=C(C=C1C(F)(F)F)NC(=N2)CCCO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 244.21
Percent Purity 97%
Chemical Name or Material 3-[5-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl]propan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.