missing translation for 'onlineSavingsMsg'
Learn More

3,6,9-Trioxaundecane-1,11-diyl-bismethanethiosulfonate, TRC

Product Code. 30457135
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457135 10 mg 10mg
30456284 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457135 Supplier Toronto Research Chemicals Supplier No. TRCT80650010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212262-02-7
Molecular Formula C9 H20 O8 S2
Molecular Weight (g/mol) 320.38
IUPAC Name 2-[2-[2-(2-methylsulfonylethoxy)ethoxy]ethoxy]ethanesulfonic acid
SMILES CS(=O)(=O)CCOCCOCCOCCS(=O)(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H20O8S2/c1-18(10,11)8-6-16-4-2-15-3-5-17-7-9-19(12,13)14/h2-9H2,1H3,(H,12,13,14)
Formula Weight 320.06
Chemical Name or Material 3,6,9-Trioxaundecane-1,11-diyl-bismethanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.