missing translation for 'onlineSavingsMsg'
Learn More

3,6-Di-2-pyridyl-1,2,4,5-tetrazine, 96%

Product Code. 15430358
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430358 25 g 25g
15410358 1 g 1g
15420358 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430358 Supplier Thermo Scientific Alfa Aesar Supplier No. H53484.14
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1671-87-0 | C12H8N6 | 236.24 g/mol

3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1671-87-0
Molecular Formula C12H8N6
Molecular Weight (g/mol) 236.24
MDL Number MFCD00121717
InChI Key JFBIRMIEJBPDTQ-UHFFFAOYSA-N
Synonym 3,6-di pyridin-2-yl-1,2,4,5-tetrazine, 3,6-di 2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridinyl-1,2,4,5-tetrazine, 3,6-bis 2-pyridyl-1,2,4,5-tetrazine, bis pyridin-2-yl-1,2,4,5-tetrazine, 3,6-bis 2-pyridinyl-1,2,4,5-tetrazine, bptz, acmc-209dvt, 3,6-di-2-pyridyl-s-tetrazine
PubChem CID 315109
IUPAC Name 3,6-dipyridin-2-yl-1,2,4,5-tetrazine
SMILES C1=CC=C(N=C1)C1=NN=C(N=N1)C1=CC=CC=N1

Specifications

Melting Point 225°C (decomposition)
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 236.24
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Di-2-pyridyl-1,2,4,5-tetrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.