missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromopyridazine, 95%

Product Code. 15487728
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487728 1 g 1g
15497728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Se returpolicy
Artikelnummer. 15487728 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H61223.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 17973-86-3 | C4H2Br2N2 | 237.882 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 17973-86-3
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.882
MDL Number MFCD00233947
InChI Key VQAFMTSSCUETHA-UHFFFAOYSA-N
PubChem CID 248852
IUPAC Name 3,6-dibromopyridazine
SMILES C1=CC(=NN=C1Br)Br

Tekniske data

Melting Point 117°C to 121°C
Color White to Yellow
Quantity 1 g
Synonym 3,6-dibromopyridazide, pyridazine, 3,6-dibromo, 3,6-dibromo pyridazine, pubchem12385, 3,6-dibromo-pyridazine, pyridazine,3,6-dibromo, 3,6-dibromo-1,2-diazine, 3,6-dibromopyridazine, synquest 3h32-9-t3
Formula Weight 237.88
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3,6-Dibromopyridazine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.