missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloropyridazine, 97%

Product Code. 10676934 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10676934 500 g Plastic bottle 500g
10030740 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10676934 Supplier Thermo Scientific Acros Supplier No. 113715000
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 141-30-0 | C4H2Cl2N2 | 148.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-30-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.98
MDL Number MFCD00006466
InChI Key GUSWJGOYDXFJSI-UHFFFAOYSA-N
Synonym pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine
PubChem CID 67331
IUPAC Name 3,6-dichloropyridazine
SMILES C1=CC(=NN=C1Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 65°C to 69°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Formula Weight 148.98
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 6-Dichloropyridazine
Show More Show Less
compliance-icons
Product Identifier
  • 3,6-Dichloropyridazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.