missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate, 95%, Thermo Scientific™

Product Code. 10743512
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10743512 1 g Amber Glass Bottle 1g
10311610 5 g Amber Glass Bottle 5g
10505470 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10743512 Supplier Thermo Scientific Maybridge Supplier No. MO08368DA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 188975-30-6
Molecular Formula C6H7F3O4S
Molecular Weight (g/mol) 232.173
InChI Key QXCAHEXKUMUTRF-UHFFFAOYSA-N
Synonym 3,6-dihydro-2h-pyran-4-yl trifluoro-methanesulfonate, 5,6-dihydro-4-trifluoromethylsulfonyloxy-2h-pyran, 3,6-dihydro-2h-pyran-4-yltrifluoromethanesulfonate, 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-pyran, 2h-5,6-dihydropyran-4-yl trifluoromethyl sulfonate, 4-trifluoromethylsulfonyloxy-5,6-dihydro-2h-pyran, trifluoro-methanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, methanesulfonic acid, 1,1,1-trifluoro-, 3,6-dihydro-2h-pyran-4-yl ester
PubChem CID 11356703
IUPAC Name 3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
SMILES C1COCC=C1OS(=O)(=O)C(F)(F)F

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 232.18
Chemical Name or Material 3, 6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.