missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Product Code. 11985761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11985761 5 g 5g
11975761 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985761 Supplier Thermo Scientific Acros Supplier No. 439750050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181180-42-7
Molecular Formula C6H7F3O3S2
Molecular Weight (g/mol) 248.24
InChI Key ZMDCEXLCDOPKGH-UHFFFAOYSA-N
Synonym 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 2,3dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 3,6-dihydro-2h-thiopyran-4-yltrifluoromethanesulfonate, trifluoro-methanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, methanesulfonic acid, trifluoro-, 3,6-dihydro-2h-thiopyran-4-yl ester
PubChem CID 22625751
IUPAC Name 3,6-dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
SMILES C1CSCC=C1OS(=O)(=O)C(F)(F)F

Specifications

Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 248.24
Percent Purity 97%
Chemical Name or Material 3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
compliance-icons compliance-icons
Product Identifier
  • 3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.