missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran

Product Code. 11985881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985881 1 g Glass bottle 1g
11955881 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985881 Supplier Thermo Scientific Acros Supplier No. 429610010
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 862129-81-5 | C11H19BO2S | 226.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862129-81-5
Molecular Formula C11H19BO2S
Molecular Weight (g/mol) 226.14
MDL Number MFCD10700154
InChI Key QZVRTORVCKGPPY-UHFFFAOYSA-N
Synonym 2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-2h-thiopyran-4-ylboronic acid pinacol ester, 2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2h-thiopyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-dihydro-2h-thiopyran-4-yl-4, 3,6-dihydrothiopyran-4-boronic acid pinacol ester, 3,6-dihydro-2h-thiopyran-4-ylboronicacidpinacolester, 3,6-dihydro-2h-thiopyran-4-yl boronic acid pinacol ester, 2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5,-tetramethyl-1,3,2-dioxaborolane
PubChem CID 46739735
IUPAC Name 2-(3,6-dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCSCC1

Spezifikation

Melting Point 42.0°C to 44.5°C
Packaging Glass bottle
Quantity 1 g
Formula Weight 226.15
Chemical Name or Material 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran
compliance-icons
Produktidentifikator
  • 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.