missing translation for 'onlineSavingsMsg'
Learn More

3,6-Diiodo-1H-indazole, TRC

Product Code. 30381157
Change view
Click to view available options
Quantity:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381157 2.5 g
30376122 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381157 Supplier Toronto Research Chemicals Supplier No. TRCD4549002.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319472-78-1
Molecular Formula C7H4I2N2
Molecular Weight (g/mol) 369.93
Synonym 1H-Indazole, 3,6-diiodo- (9CI, ACI), 3,6-Diiodo-1H-indazole (ACI), 3,6-Diiodo-2H-indazole
SMILES Ic1ccc2c(I)n[nH]c2c1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H4I2N2/c8-4-1-2-5-6(3-4)10-11-7(5)9/h1-3H,(H,10,11)
Chemical Name or Material 3,6-Diiodo-1H-indazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.