missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dithia-1,8-octanediol, 97%

Product Code. 11433228
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433228 10 g 10g
11443228 50 g 50g
11453228 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433228 Supplier Thermo Scientific Alfa Aesar Supplier No. A17654.09
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5244-34-8 | C6H14O2S2 | 182.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5244-34-8
Molecular Formula C6H14O2S2
Molecular Weight (g/mol) 182.296
MDL Number MFCD00002911
InChI Key PDHFSBXFZGYBIP-UHFFFAOYSA-N
Synonym 3,6-dithia-1,8-octanediol, 1,2-bis 2-hydroxyethylthio ethane, 3,6-dithiaoctane-1,8-diol, dithiaoctanediol, ethylenedithioethanol, ethanol, 2,2'-1,2-ethanediylbis thio bis, unii-kbc80263g6, 2,2'-ethane-1,2-diylbis sulfanediyl diethanol, 2,2'-ethylenedithio diethanol, ethanol, 2,2'-ethylenedithio di
PubChem CID 78904
IUPAC Name 2-[2-(2-hydroxyethylsulfanyl)ethylsulfanyl]ethanol
SMILES C(CSCCSCCO)O

Specifications

Melting Point 63°C to 67°C
Boiling Point 170°C (0.8 mmHg)
Quantity 10 g
Beilstein 1739193
Formula Weight 182.31
Percent Purity 97%
Chemical Name or Material 3,6-Dithia-1,8-octanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.