missing translation for 'onlineSavingsMsg'
Learn More

3,7-Diaminophenothiazin-5-ium acetate, BLD Pharm™

Product Code. 31449814
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31449814 5 g
31443983 1 g
31448885 25 g
31445528 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31449814 Supplier BLD Pharm Supplier No. BD1212375g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dye content 85 %

Specifications

CAS 78338-22-4
Molecular Formula C14H13N3O2S
MDL Number MFCD00081194
Quantity 5 g
SMILES O=C(C)[O-].NC1=CC=C2N=C3C=CC(N)=CC3=[S+]C2=C1
Molecular Weight (g/mol) 287.34
Chemical Name or Material 3,7-Diaminophenothiazin-5-ium acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.