missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetylaminotoluene, TRC
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
missing translation for 'unitSize'
10g
Spezifikation
Spezifikation
| Chemical Name or Material | 3-Acetylaminotoluene |
| CAS | 537-92-8 |
| Molecular Formula | C9 H11 N O |
| Quantity | 10 g |
| Synonym | Acetamide, N-(3-methylphenyl)-, m-Acetotoluidide (7CI,8CI), N-(3-Methylphenyl)acetamide, 1-Acetamido-3-methylbenzene, 3-Acetamidotoluene, 3-Methylacetanilide, 3'-Methylacetanilide, Aceto-m-aminotoluene, N-Acetyl-3-methylaniline, N-Acetyl-m-toluidine, N-m-Tolylacetamide, NSC 3103, m-Acetotoluide, m-Methylacetanilide, m-N-Acetyltoluidine, m-Tolylacetamide |
| InChI Formula | InChI=1S/C9H11NO/c1-7-4-3-5-9(6-7)10-8(2)11/h3-6H,1-2H3,(H,10,11) |
| SMILES | CC(=O)Nc1cccc(C)c1 |
| IUPAC Name | N-(3-methylphenyl)acetamide |
| Molecular Weight (g/mol) | 149.19 |
| Formula Weight | 149.0841 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction