missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyldeoxynivalenol, TRC

Product Code. 18013720
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18013720 5 mg 5mg
18082930 1 mg 1mg
18090910 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18013720 Supplier Toronto Research Chemicals Supplier No. TRCA1735055MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50722-38-8
Molecular Formula C17 H22 O7
Molecular Weight (g/mol) 338.35
Synonym (3α,7α)-3-(Acetyloxy)-12,13-epoxy-7,15-dihydroxytrichothec-9-en-8-one, Spiro[2,5-methano-1-benzoxepin-10,2'-oxirane], trichothec-9-en-8-one deriv., 3-Acetyldeoxynivalenol, 3α-Acetoxy-12,13-epoxytrichothec-9-en-8-one, DON 3-acetate, Dehydronivalenol monoacetate, Deoxynivalenol 3-acetate, Deoxynivalenol monoacetate, Monoacetyldeoxynivalenol, NSC 267036
IUPAC Name [(1R,2R,3S,7R,9R,10R,12S)-3-hydroxy-2-(hydroxymethyl)-1,5-dimethyl-4-oxospiro[8-oxatricyclo[7.2.1.02,7]dodec-5-ene-12,2'-oxirane]-10-yl] acetate
SMILES CC(=O)O[C@H]1C[C@]2(C)C3(CO3)[C@H]1O[C@H]4C=C(C)C(=O)[C@H](O)[C@@]24CO

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H22O7/c1-8-4-11-16(6-18,13(21)12(8)20)15(3)5-10(23-9(2)19)14(24-11)17(15)7-22-17/h4,10-11,13-14,18,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17?/m0/s1
Formula Weight 338.1366
Chemical Name or Material 3-Acetyl-deoxynivalenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.