missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylphenylboronic acid, 96%, Thermo Scientific™

Product Code. 10497582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10497582 1 g Glass bottle 1g
10703023 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10497582 Supplier Thermo Scientific Acros Supplier No. 358830010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 204841-19-0
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD01074678
InChI Key SJGGDZCTGBKBCK-UHFFFAOYSA-N
Synonym 3-acetylphenyl boronic acid, 3-acetylbenzeneboronic acid, 3-acetylphenylboronicacid, m-acetylphenylboronic acid, 3'-boronoacetophenone, boronic acid, 3-acetylphenyl, 1-3-dihydroxyboranyl phenyl ethan-1-one, pubchem2751, acmc-209faq, 3-acetylphenyboronic acid
PubChem CID 2734310
ChEBI CHEBI:86552
IUPAC Name (3-acetylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)C)(O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 212°C to 214°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.97
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Acetylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.