missing translation for 'onlineSavingsMsg'
Learn More

3-Alpha-Hydroxy Pravastatin Sodium Salt, TRC

Product Code. 17950505
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950505 10 mg 10mg
17983205 1 mg 1mg
17973005 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17950505 Supplier Toronto Research Chemicals Supplier No. TRCH95231010MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81093-43-8
Molecular Formula C23 H35 O7 . Na
Molecular Weight (g/mol) 446.51
Synonym (3R,5R)-3,5-Dihydroxy-7-[(1S,2R,3S,8S,8aR)-3-hydroxy-2-methyl-8-[[(2S)-2-methylbutanoyl]oxy]-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] heptanoate, 1-Naphthaleneheptanoic acid, 1,2,3,7,8,8a-hexahydro-β,δ,3-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-, sodium salt (1:1), (βR,δR,1S,2R,3S,8S,8aR)-, 1-Naphthaleneheptanoic acid, 1,2,3,7,8,8a-hexahydro-β,δ,3-trihydroxy-2-methyl-8-(2-methyl-1-oxobutoxy)-, monosodium salt, [1S-[1α(βS*,δS*),2α,3β,8β(R*),8aα]]- (9CI), Sodium (βR,δR,1S,2R,3S,8S,8aR)-1,2,3,7,8,8a-hexahydro-β,δ,3-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoate, Pravastatin Related Compound A (USP), 3α-Hydroxy Pravastatin sodium salt, 3α-Hydroxypravastatin sodium salt
IUPAC Name sodium;(3R,5R)-7-[(1S,2R,3S,8S,8aR)-3-hydroxy-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,3,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
SMILES [Na+].CC[C@H](C)C(=O)O[C@H]1CC=CC2=C[C@@H](O)[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H36O7.Na/c1-4-13(2)23(29)30-20-7-5-6-15-10-19(26)14(3)18(22(15)20)9-8-16(24)11-17(25)12-21(27)28;/h5-6,10,13-14,16-20,22,24-26H,4,7-9,11-12H2,1-3H3,(H,27,28);/q;+1/p-1/t13-,14+,16+,17+,18-,19+,20-,22-;/m0./s1
Formula Weight 446.228
Percent Purity >95%
Chemical Name or Material 3a-Hydroxy Pravastatin Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.