missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-(4-chlorophenyl)-1H-pyrazole, 95%, Thermo Scientific™

Product Code. 15424957
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424957 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15424957 Supplier Thermo Scientific Alfa Aesar Supplier No. H32252.MD
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 66000-39-3
Melting Point 107°C to 108°C
Molecular Formula C9H8ClN3
MDL Number MFCD09027932
Quantity 250 mg
Synonym 1-4-chlorophenyl-1h-pyrazol-3-amine, 3-amino-1-4-chlorophenyl-1h-pyrazole, 1-4-chlorophenyl pyrazol-3-amine, 1-p-chlorophenyl-3-amino-pyrazole, 1-4-chlorophenyl-1h-pyrazol-3-ylamine, 1-4-chloro-phenyl-1h-pyrazol-3-ylamine
InChI Key SHCVEWFQFUKSDP-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N2C=CC(=N2)N)Cl
IUPAC Name 1-(4-chlorophenyl)pyrazol-3-amine
Molecular Weight (g/mol) 193.634
PubChem CID 13626945
Formula Weight 193.64
Percent Purity 95%
Chemical Name or Material 3-Amino-1-(4-chlorophenyl)-1H-pyrazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.