missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-bromoisoquinoline, 97+%

Product Code. 11375349
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11375349 1 g 1g
11385349 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11375349

Brand: Thermo Scientific Alfa Aesar H50368.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13130-79-5 | C9H7BrN2 | 223.073 g/mol

3-Amino-1-bromoisoquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1-bromoisoquinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13130-79-5
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD00102193
InChI Key PSQUIUNIVDKHJK-UHFFFAOYSA-N
Synonym 3-amino-1-bromoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromoisoquinolin-3-ylamine, 1-bromo-3-isoquinolylamine, 3-amino-1-bromo-isoquinoline, 1-bromoisoquinoline-3-amine, acmc-1by0f, 1-bromo-3-aminoisoquinoline, 1-bromanylisoquinolin-3-amine, 3-isoquinolinamine, 1-bromo
PubChem CID 289845
IUPAC Name 1-bromoisoquinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(N=C2Br)N

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 223.07
Percent Purity ≥97%
Chemical Name or Material 3-Amino-1-bromoisoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.