missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanol, 99%

Product Code. 10202122 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10202122 5 mL Glass bottle 5mL
10794781 500 mL Glass bottle 500mL
10616204 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10202122 Supplier Thermo Scientific Acros Supplier No. 104460050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 156-87-6 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-87-6
MDL Number MFCD00008223
InChI Key WUGQZFFCHPXWKQ-UHFFFAOYSA-N
Synonym 3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
PubChem CID 9086
IUPAC Name 3-aminopropan-1-ol
SMILES C(CN)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 10.0°C to 12.0°C
Color Colorless to Yellow
Density 0.9820g/mL
Boiling Point 187.0°C to 188.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C3H9NO
Linear Formula H2N(CH2)3OH
Refractive Index 1.4595 to 1.4615
Quantity 5 mL
Beilstein 04, 288
Specific Gravity 0.982
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohols,esters, chloroform, acetone, slightly soluble in arom. and aliph. hydrocarbons
Molecular Weight (g/mol) 75.11
Viscosity 36 mPa.s (20°C)
Formula Weight 75.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Amino-1-propanol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Amino-1-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.