missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-pyrazole-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 11420188
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420188 5 g 5g
11430188 0.25 g 0.25g
11410188 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420188 Supplier Thermo Scientific Alfa Aesar Supplier No. B23751.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41680-34-6 | Molecular Formula: C4H5N3O2 | Molecular Weight: 127.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 41680-34-6
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00005239
InChI Key KMRVTZLKQPFHFS-UHFFFAOYSA-N
Synonym 3-amino-1h-pyrazole-4-carboxylic acid, 3-aminopyrazole-4-carboxylic acid, 3-amino-4-pyrazolecarboxylic acid, 3-amino-4-carboxypyrazole, 1h-pyrazole-4-carboxylic acid, 3-amino, 4-pyrazolecarboxylic acid, 3-amino, pyrazole-4-carboxylic acid, 3-amino, 3-aminopyrazole-4-carboxylicacid, 3-amino-4-pyrazole carboxylic acid, 5-aminopyrazole-4-carboxylic acid
PubChem CID 96774
IUPAC Name 5-amino-1H-pyrazole-4-carboxylic acid
SMILES C1=NNC(=C1C(=O)O)N

Spezifikation

Melting Point ∼125°C (decomposition)
Quantity 5 g
Formula Weight 127.1
Percent Purity 95%
Chemical Name or Material 3-Amino-1H-pyrazole-4-carboxylic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.